Barium hydroksid oktahydrat, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

1.01735.1000
1.01735.1000EA 1880 NOK
1.01735.1000
Barium hydroksid oktahydrat, Supelco®
Barium hydroksid oktahydrat
Formel: Ba(OH)₂·8H₂O
Kokepunkt: 780 °C (1013 hPa)
Smeltepunkt: 78 °C
Tetthet: 2,18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149152
CAS-nummer: 12230-71-6
EINECS: 241-234-5
FN: 2923
ADR: 8 (6.1),II
Merck Indeks: 13,00980

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR