Testsett, Syrekapasitet til pH 4,3 (total alkalitet) for Spectroquant®, Supelco®
Leverandør: Merck
1.01758.0001
1.01758.0001EA 2975 NOK
1.01758.0001
Testsett, Syrekapasitet til pH 4,3 (total alkalitet) for Spectroquant®, Supelco®
Miljø testkit
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used, not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All necessary reagents for the analysis are contained in the cells for most test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.