Petroleumbensin, 100…120 °C, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.01781.1000
1.01781.1000EA 813 NOK
1.01781.1000
Petroleumbensin, 100…120 °C, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Petroleumbensin
Kokepunkt: 100…120 °C (1013 hPa)
Tetthet: 0,7 g/cm³ (20 °C)
Flash punkt: –10 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081849
CAS-nummer: 64742-49-0
EINECS: 265-151-9
FN: 1268
ADR: 3,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR