Saltsyre 37%, EMPARTA® ACS analytical reagent

Leverandør: Merck
Danger

Synonymer: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ200

1.01834.2011
1.01834.2011EA 304.5 NOK
1.01834.2011
Saltsyre 37%, EMPARTA® ACS analytical reagent
Saltsyre
Formel: HCl
MW: 36.46 g/mol
Smeltepunkt: –30 °C
Tetthet: 1.18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -28 °C
Boiling Point 45 °C
Vapor Pressure 190 hPa (20 °C)
Density 1.19 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR