Dimetylsulfoksid, SupraSolv® for headspace gas chromatography, Supelco®

Leverandør: Merck

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

M-Clarity™ quality level = MQ100

1.01900.1000 1.01900.2500 1.01900.0500
1.01900.1000EA 1036.97 NOK
1.01900.1000 1.01900.2500 1.01900.0500
Dimetylsulfoksid, SupraSolv® for headspace gas chromatography, Supelco®
Dimetylsulfoksid

SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual
solvents in drug substances, excipients and drug products according to ICH, Ph. Eur. and USP - SupraSolv® headspace solvents are tested for all residual solvents defined by the ICH.

Formel: (CH₃)₂SO
Kokepunkt: 189 °C (1013 hPa)
Tetthet: 1,101 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 1000 g/L (20 °C)
Molar Mass 18.4 °C
Bulk Density 78.1 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0%(V)
Density 1.10 g/cm³ (20 °C)
Ignition Point 301 °C
Saturation Concentration 8.0 g/m³ (20 °C)
Viscosity Kinematic 2.14 mm²/s


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR