Brom, Suprapur® for trace analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.01947.0250
1.01947.0250EA 6630 NOK
1.01947.0250
Brom, Suprapur® for trace analysis, Supelco®
Bromine
Formel: Br₂
MW: 159.81 g/mol
Kokepunkt: 58.7
Smeltepunkt: –7.2 °C
Tetthet: 3.12 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010896
CAS-nummer: 7726-95-6
EINECS: 231-778-1
FN: 1744
ADR: 8 (6.1) I
Merck Indeks: 13,01378

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR