tert-Butylmetyleter, Uvasol® for spectroscopy, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.01984.1000
1.01984.1000EA 801 NOK
1.01984.1000
tert-Butylmetyleter, Uvasol® for spectroscopy, Supelco®
tert-Butylmetyleter
Formel: (CH₃)₃COCH₃
Kokepunkt: 55,2 °C (1013 hPa)
Tetthet: 0,7405 g/cm³ (20 °C)
Flash punkt: –28 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008812
CAS-nummer: 1634-04-4
EINECS: 216-653-1
FN: 2398
ADR: 3,II
Merck Indeks: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 42 g/L (20 °C)
Melting Point -108.6 °C
Molar Mass 88.15 g/mol
Boiling Point 55.3 °C (1013 hPa)
Vapor Pressure 268 hPa (20 °C)
Flash Point -28 °C
Explosion Limit 1.5 - 8.5%(V)
Density 0.74 g/cm³ (20 °C)
pH (H₂O) not applicable
Ignition Point 460 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR