Kadmium, coarse powder, EMSURE® ∼ 0.3-1.6 mm for analysis, suitable for filling of reductors, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.02001.0250 1.02001.1000
1.02001.0250EA 0 NOK
1.02001.0250 1.02001.1000
Kadmium, coarse powder, EMSURE® ∼ 0.3-1.6 mm for analysis, suitable for filling of reductors, Supelco®
Kadmium
Formel: Cd
Kokepunkt: 767 °C (1013 hPa)
Smeltepunkt: 321 °C
Tetthet: 8,6 g/cm3 (22 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010914
CAS-nummer: 7440-43-9
EINECS: 231-152-8
FN: 3288
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.001 %
Pb (Lead) ≤ 0.01 %
Zn (Zinc) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR