Kadmiumacetat dihydrat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger

Synonymer: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate

M-Clarity™ quality level = MQ300

1.02003.0500
1.02003.0500EA 2320 NOK
1.02003.0500
Kadmiumacetat dihydrat, EMSURE® for analysis, Supelco®
Kadmiumacetat dihydrat
Formel: (H₃CCOO)₂Cd·2H₂O
Kokepunkt: 105…107 °C (4 mmHg)
Smeltepunkt: 256 °C
Tetthet: 2,341 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150020
CAS-nummer: 5743-04-4
EINECS: 208-853-2
FN: 2570
ADR: 6.1,II
Merck Indeks: 13,01614

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 256 °C (anhydrous)
Molar Mass 266.52 g/mol
Bulk Density 950 kg/m3
Density 2.01 g/cm3 (20 °C)
pH 7 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR