Kadmiumoksid, Supelco®

Leverandør: Merck
Danger

Synonymer: Cadmium (II) oxide

M-Clarity™ quality level = MQ200

1.02015.5000
1.02015.5000EA 7290 NOK
1.02015.5000
Kadmiumoksid, Supelco®
Kadmiumoksid
Formel: CdO
MW: 128.41 g/mol
Kokepunkt: 1559
Smeltepunkt: ∼1426 °C
Tetthet: 8.15 g/cm³ (20 °C)
MDL nummer: MFCD00010921
CAS-nummer: 1306-19-0
EINECS: 215-146-2
FN: 2570
ADR: 6.1 I
Merck Indeks: 13,01623

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR