Kadmium(II)sulfat 8/3-hydrat, EMSURE® ACS for analysis

Leverandør: Merck
Danger

Synonymer: Cadmium sulphate, 8/3-hydrate, Cadmium sulphate octahydrate

M-Clarity™ quality level = MQ300

1.02027.0100
1.02027.0100EA 974 NOK
1.02027.0100
Kadmium(II)sulfat 8/3-hydrat, EMSURE® ACS for analysis
Kadmium(II)sulfat 8/3-hydrat
Formel: 3CdSO₄·8H₂O
MW: 256.51 g/mol (anhy)
Smeltepunkt: 41
Tetthet: 3.79 g/cm³ (20 °C)
MDL nummer: MFCD02253000
CAS-nummer: 7790-84-3
EINECS: 233-331-6
FN: 2570
ADR: 6.1 III
Merck Indeks: 13,01627

Bestill nå

Spesifikasjon Testresultater

Assay (complexometric, CdSO₄.8/3 H₂O) 98.0 - 102.0 %
Insoluble matter ≤ 0.005 %
pH-value (5 %; water) 3.0 - 6.0
Chloride (Cl) ≤ 0.001 %
Total nitrogen (N) ≤ 0.0005 %
Ca (Calcium) ≤ 0.005 %
Cu (Copper) ≤ 0.0005 %
Fe (Iron) ≤ 0.0005 %
K (Potassium) ≤ 0.01 %
Na (Sodium) ≤ 0.005 %
Pb (Lead) ≤ 0.002 %
Zn (Zinc) ≤ 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR