Dragendorff's reagent, spray reagent for TLC, Supelco®
Leverandør: Merck
Danger
M-Clarity™ quality level = MQ100
1.02035.0100
1.02035.0100EA 660 NOK
1.02035.0100
Dragendorff's reagent, spray reagent for TLC, Supelco®
Dragendorff's reagent
Kokepunkt: 77
Tetthet: 1.2 g/cm³ (20 °C)
Flash punkt: –4 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00283106
CAS-nummer: 39775-75-2
FN: 2924
ADR: 3 (8) II
Bestill nå
Spesifikasjon Testresultater
Boiling Point 77 °C
Density 0.95 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid