Kalsiumhydroksid, Supelco®

Leverandør: Merck
Danger

Synonymer: Slaked lime, Calcium dihydroxide

M-Clarity™ quality level = MQ100

1.02111.0025
1.02111.0025EA 1720 NOK
1.02111.0025
Kalsiumhydroksid, Supelco®
Kalsiumhydroksid
Formel: Ca(OH)₂
Smeltepunkt: 550 °C
Tetthet: 2,24 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010901
CAS-nummer: 1305-62-0
EINECS: 215-137-3
FN: 3262
ADR: 8,III
Merck Indeks: 13,01675

Bestill nå

Spesifikasjon Testresultater

Solubility 1.7 g/l (20 °C)
Melting Point 550 °C (decomposition)
Molar Mass 74.09 g/mol
Bulk Density 400 kg/m3
Boiling Point 2850 °C (1013 hPa) (decomposition)
Vapor Pressure (20 °C)
Density 2.24 g/cm3 (20 °C)
pH 12.6 (H2O, 20 °C) (saturated solution)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR