Karbondisulfid Reag. Ph. Eur., ACS analytical reagent, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.02214.1000
1.02214.1000EA 2050 NOK
1.02214.1000
Karbondisulfid Reag. Ph. Eur., ACS analytical reagent, Supelco®
Karbondisulfid
Formel: CS₂
MW: 76.14 g/mol
Kokepunkt: 46.2
Smeltepunkt: –111.6 °C
Tetthet: 1.2632 g/cm³ (20 °C)
Flash punkt: –30 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011321
CAS-nummer: 75-15-0
EINECS: 200-843-6
FN: 1131
ADR: 3 (6.1) I
Merck Indeks: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 2.1 g/l (20 °C)
Melting Point -111.6 °C
Molar Mass 76.13 g/mol
Boiling Point 46.5 °C (1013 hPa)
Vapor Pressure 398 hPa (20 °C)
Flash Point -30 °C
Explosion Limit 1 - 60 %(V)
Density 1.26 g/cm3 (20 °C)
Ignition Point 95 °C
Evaporation number 1.8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR