Ethidium bromide adsorber for decontamination of ethidium bromide from staining solutions, Sigma-Aldrich®
Leverandør: Merck
1.02243.0001
1.02243.0001EA 1190 NOK
1.02243.0001
Ethidium bromide adsorber for decontamination of ethidium bromide from staining solutions, Sigma-Aldrich®
Elektroforese reagenser Reagenser for fjerning av fargeløsning
Developed specifically for the safe and simple removal of ethidium bromide (EtBr) from aqueous staining solutions and running buffers used in nucleic acid separation gels.
  • Form: Gray-brown suspension
Typical concentrations in these solutions are 0.5 - 10 mg/L. EtBr is bound and concentrated by the adsorber column, allowing removal of the filtrate. Each column normally binds at least 300 mg of ethidium bromide from TAE or TBE buffers. Residual capacity is easily visualized because the EtBr appears as a dark red to black band on the column. As long as this band has not reached the bottom of the column bed, the concentration of dye is still below the column capacity.
Special Instructions: When the adsorber column is not in use, both ends should be closed off. The column should not be allowed to dry out, as air bubbles may form in the column, hindering flow rate.