Ammoniumcerium(IV)nitrat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.02276.0100 1.02276.1000
1.02276.0100EA 1950 NOK
1.02276.0100 1.02276.1000
Ammoniumcerium(IV)nitrat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Ammoniumcerium(IV)nitrat
Formel: (NH₄)₂Ce(NO₃)₆
Smeltepunkt: 107…108 °C
Tetthet: 2,2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00151121
CAS-nummer: 16774-21-3
EINECS: 240-827-6
FN: 3085
ADR: 5.1 (8),II
Merck Indeks: 13,00509

Bestill nå

Spesifikasjon Testresultater

Solubility 1410 g/l (20 °C)
Melting Point 108 °C
Molar Mass 548.22 g/mol
Bulk Density 1200 kg/m3
Density 2.49 g/cm3
pH 1 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR