Kalsiumklorid tetrahydrat, Suprapur® for trace analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ100

1.02384.0100
1.02384.0100EA 3880 NOK
1.02384.0100
Kalsiumklorid tetrahydrat, Suprapur® for trace analysis, Supelco®
Kalsiumklorid tetrahydrat
Formel: CaCl₂·4H₂O
Kokepunkt: ∼1600 °C (1013 hPa)
Smeltepunkt: 772 °C
Tetthet: 1,71 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
CAS-nummer: 25094-02-4
EINECS: 233-140-8

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 772 °C (anhydrous substance)
Molar Mass 183.05 g/mol
pH 5 - 7 (50 g/l, H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR