di-Natriumoksalat, Certipur®, Supelco®

Leverandør: Merck
Warning

Synonymer: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

M-Clarity™ quality level = MQ100

1.02407.0060
1.02407.0060EA 1220 NOK
1.02407.0060
di-Natriumoksalat, Certipur®, Supelco®
di-Natriumoksalat
Formel: Na₂C₂O₄
Smeltepunkt: 250…270 °C
Tetthet: 2,27 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012465
CAS-nummer: 62-76-0
EINECS: 200-550-3
Merck Indeks: 13,08723

Bestill nå

Spesifikasjon Testresultater

Solubility 37 g/l (20 °C)
Melting Point 250 - 270 °C (decomposition)
Molar Mass 134 g/mol
Bulk Density 600 kg/m3
Density 2.27 g/cm3 (20 °C)
pH 8 (30 g/l, H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR