kobolt (II) acetat tetrahydrat

Leverandør: Merck
Danger

1.02530.0250
1.02530.0250EA 1150 NOK
1.02530.0250
kobolt (II) acetat tetrahydrat
kobolt (II) acetat tetrahydrat
Formel: (H₃CCOO)₂Co·4H₂O
Smeltepunkt: 140 °C
Tetthet: 1,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149569
CAS-nummer: 6147-53-1
EINECS: 200-755-8
FN: 3077
ADR: 9,III
Merck Indeks: 12,02494

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR