Kobalt(II)nitrat heksahydrat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.02536.0100 1.02536.0250
1.02536.0100EA 1880 NOK
1.02536.0100 1.02536.0250
Kobalt(II)nitrat heksahydrat, EMSURE® for analysis, Supelco®
Kobalt(II)nitrat heksahydrat
Formel: Co(NO₃)₂·6H₂O
Smeltepunkt: 57 °C
Tetthet: 1,88 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149647
CAS-nummer: 10026-22-9
EINECS: 233-402-1
FN: 3077
ADR: 9,II
Merck Indeks: 13,02469

Bestill nå

Spesifikasjon Testresultater

Solubility 2170 g/l (100 °C)
Melting Point 57 °C
Molar Mass 291.04 g/mol
Bulk Density 800 kg/m3
Density 1.87 g/cm3 (20 °C)
pH 4.0 (100 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR