Kobalt(II)klorid heksahydrat, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Leverandør: Merck
Danger

Synonymer: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

M-Clarity™ quality level = MQ300

1.02539.0100 1.02539.0250
1.02539.0100EA 3480 NOK
1.02539.0100 1.02539.0250
Kobalt(II)klorid heksahydrat, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Kobalt(II)klorid heksahydrat
Formel: CoCl₂·6H₂O
Kokepunkt: 110 °C (1013 hPa)
Smeltepunkt: 56 °C
Tetthet: 1,92 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149652
CAS-nummer: 7791-13-1
EINECS: 231-589-4
FN: 3077
ADR: 9,III
Merck Indeks: 13,02462

Bestill nå

Spesifikasjon Testresultater

Solubility 191 g/l (100 °C)
Melting Point 56 °C
Molar Mass 237.90 g/mol
Bulk Density 1250 kg/m3
Density 1.92 g/cm3 (25 °C)
pH 4.9 (50 g/l, H2O, 25 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR