Schiff reagent intense, Sigma-Aldrich®
Leverandør: Merck
Warning
1.02572.1000
1.02572.1000EA 1125 NOK
1.02572.1000
Schiff reagent intense, Sigma-Aldrich®
Schiff reagent
For the detection of aldehydes, polysaccharides (glycogen), and mucosubstances (glycoproteins, glycolipids, mucins)
The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for the detection of aldehyde and mucosubstances in microscopy. Schiff’s reagent intense is a ready to use, high capacity staining solution (up to 3000 slides/500 ml). It can be stored at room temperature which allows for 10 minutes of time saving per staining procedure. This new formulation provides for more intense staining results with clearer differentiation between cell types.