Kobber(II)sulfat pentahydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.02790.0250 1.02790.1000 1.02790.5000 1.02790.9050
1.02790.0250EA 990 NOK
1.02790.0250 1.02790.1000 1.02790.5000 1.02790.9050
Kobber(II)sulfat pentahydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Kobber(II)sulfat pentahydrat
Formel: CuSO₄·5H₂O
Tetthet: 2,284 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149681
CAS-nummer: 7758-99-8
EINECS: 231-847-6
FN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Indeks: 13,02682

Limited Time Sale

Select items below are currently on sale

Bestill nå

Spesifikasjon Testresultater

Assay (iodometric) 99.0-100.5%
Insoluble matter 0.005% max
Chloride (Cl) 0.0005% max
Total nitrogen (N) 0.001% max
Calcium (Ca) 0.005% max
Iron (Fe) 0.003% max
Potassium (K) 0.001% max
Sodium(Na) 0.005% max
Nickel (Ni) 0.005% max
Lead (Pb) 0.005% max
Zinc (Zn) 0.03% max
Corresponds to ACS,ISO,Reag. Ph Eur

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR