Kobber(II)sulfat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Warning

Synonymer: Cupric sulphate

M-Clarity™ quality level = MQ300

1.02791.0250 1.02791.1000
1.02791.0250EA 909 NOK
1.02791.0250 1.02791.1000
Kobber(II)sulfat, EMSURE® for analysis, Supelco®
Kobber(II)sulfat
Formel: CuSO₄
Smeltepunkt: 200 °C
Tetthet: 3,603 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010981
CAS-nummer: 7758-98-7
EINECS: 231-847-6
FN: 3077
ADR: 9,III
Merck Indeks: 12,02722

Bestill nå

Spesifikasjon Testresultater

Solubility 203 g/l (20 °C)
Molar Mass 159.61 g/mol
Bulk Density 800 kg/m3
Density 3.60 g/cm3 (20 °C)
pH 3.5 - 4.5 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR