Acetonitril-D3 (99% D) for NMR spectroscopy, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

M-Clarity™ quality level = MQ200

1.02904.0010
1.02904.0010EA 1110 NOK
1.02904.0010
Acetonitril-D3 (99% D) for NMR spectroscopy, Sigma-Aldrich®
Acetonitril-D3
Formel: D₃CCN
MW: 44.03 g/mol
Kokepunkt: 80.7
Smeltepunkt: –42 °C
Tetthet: 0.844 g/cm³ (20 °C)
Flash punkt: 2 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001881
CAS-nummer: 2206-26-0
EINECS: 218-616-5
FN: 1648
ADR: 3 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR