Dimetylsulfoksid, Uvasol® for spectroscopy, Supelco®

Leverandør: Merck

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

M-Clarity™ quality level = MQ100

1.02950.0500 1.02950.2500
1.02950.0500EA 1400 NOK
1.02950.0500 1.02950.2500
Dimetylsulfoksid, Uvasol® for spectroscopy, Supelco®
Dimetylsulfoksid

Accurate analytic results in UV/Vis and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for five typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph. Eur. and ACS.

Formel: (CH₃)₂SO
Kokepunkt: 189 °C (1013 hPa)
Tetthet: 1,101 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 1000 g/L (20 °C)
Melting Point 18.4 °C
Molar Mass 78.1 g/mol
Boiling Point 189 °C (1013 hPa)
Vapour Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0%(V)
Density 1.10 g/cm³ (20 °C)
Ignition Point 301 °C
Saturation Concentration 8.0 g/m³ (20 °C)
Viscosity Kinematic 2.14 mm²/s


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR