3,3'-Dimethylnaphthidine indicator for metal titration, Supelco®

Leverandør: Merck

Synonymer: 4,4'-Diamino-3,3'dimethyl-1,1'-binaphthyl, 3,3'-Dimethyl-[1,1'-binaphthalene]-4,4'-diamine, 4-(4-amino-3-methylnaphthalen-1-yl)-2-methylnaphthalen-1-amine

M-Clarity™ quality level = MQ100

1.03122.0001
1.03122.0001EA 6460 NOK
1.03122.0001
3,3'-Dimethylnaphthidine indicator for metal titration, Supelco®
3,3'-Dimethylnaphthidine

Application note: Method A or Method B
Method A: For the measurement the method 195, which is pre-calibrated in the Spectroquant® Pharo-and NOVA-photometers can be used. To increase the precision of the results a self-made blank according to the application is recommended generally.
Method B: For the measurement the pre-calibrated method 195 is not applicable. Instead of that, a self-made calibration according the application is to perform.

Formel: C₂₂H₂₀N₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00042706
CAS-nummer: 13138-48-2
EINECS: 236-069-0

Bestill nå

Spesifikasjon Testresultater

Appearance of solution (10 g/l; ethanol) passes test
Loss on drying (110 °C) ≤ 1.0 %
Suitability as indicator (for metal titration) passes test
Suitability for bromate determination in water and drinking water for Pharo- or NOVA-spectrophotometer passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR