Eriochrome blue black B indicator for metal titration, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

1.03168.0025
1.03168.0025EA 671 NOK
1.03168.0025
Eriochrome blue black B indicator for metal titration, Supelco®
Eriochrome blue black B
Formel: C₂₀H₁₃N₂NaO₅S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003934
CAS-nummer: 3564-14-5
EINECS: 222-639-6

Bestill nå

Spesifikasjon Testresultater

Solubility 20 g/l (20 °C)
Molar Mass 416.39 g/mol
Bulk Density 540 kg/m3
pH 9.5 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR