Absorbent for acid spills, Chemizorb® H+
Leverandør: Merck
1.03874.2000 1.03874.5000
1.03874.2000EA 1325 NOK
1.03874.2000 1.03874.5000
Absorbent for acid spills, Chemizorb® H+
Materialer for håndtering av søl
Chemizorb H+ is a granulate mixture that consists of a mineral co-polymer absorbent, an alkaline neutraliser, and a pH indicator.
  • Cover the spill with the amount necessary to neutralise and absorb
  • pH indicator makes it possible to monitor the neutralisation process
  • 100 g of Chemizorb H+ required to absorb and neutralise 100 ml of acid
The neutralisation process results in the release of CO₂. When the acid has been neutralised, the colour of the indicator changes from red to orange/yellow.