Jern(III)nitrat nonahydrat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.03883.0250 1.03883.1000
1.03883.0250EA 590 NOK
1.03883.0250 1.03883.1000
Jern(III)nitrat nonahydrat, EMSURE® for analysis, Supelco®
Jern(III)nitrat nonahydrat
Formel: Fe(NO₃)₃·9H₂O
Kokepunkt: 125 °C (1013 hPa)
Smeltepunkt: 47 °C
Tetthet: 1,68 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149708
CAS-nummer: 7782-61-8
EINECS: 233-899-5
FN: 1466
ADR: 8,II
Merck Indeks: 13,04057

Bestill nå

Spesifikasjon Testresultater

Assay (complexometric) 99.0 - 101.0 %
Insoluble matter ≤ 0.005 %
Free acid (as HNO₃) ≤ 0.3 %
Chloride (Cl) ≤ 0.0005 %
Phosphate (PO₄) ≤ 0.005 %
Sulfate (SO₄) ≤ 0.005 %
Ca (Calcium) ≤ 0.005 %
Cu (Copper) ≤ 0.005 %
K (Potassium) ≤ 0.005 %
Mg (Magnesium) ≤ 0.001 %
Mn (Manganese) ≤ 0.02 %
Na (Sodium) ≤ 0.005 %
Pb (Lead) ≤ 0.001 %
Zn (Zinc) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR