Glykolsyre, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger

Synonymer: 2-Hydroxy acid, 2-Hydroxyacetic acid

M-Clarity™ quality level = MQ300

1.04106.0100
1.04106.0100EA 2300 NOK
1.04106.0100
Glykolsyre, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Glykolsyre
Formel: C₂H₄O₃
Kokepunkt: 266 °C (1013 hPa)
Smeltepunkt: 79…80 °C
Tetthet: 1,49 g/cm³ (20 °C)
Flash punkt: > 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004312
CAS-nummer: 79-14-1
EINECS: 201-180-5
FN: 3261
ADR: 8,II
Merck Indeks: 13,04511

Bestill nå

Spesifikasjon Testresultater

Assay (acidimetric) ≥ 99.5 %
Identity (IR) passes test
Melting range (lower value) ≥ 75 °C
Melting range (upper value) ≤ 79 °C
Heavy metals (as Pb) ≤ 10 ppm
Iron (Fe) ≤ 5 ppm
Water (K. F.) ≤ 0.50 %
Density 1.49 g/cm3 (25 °C)
pH 2 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR