n-Heksan, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.04374.1000 1.04374.2500 1.04374.6025 1.04374.2511 1.04374.4000
1.04374.1000EA 898 NOK
1.04374.1000 1.04374.2500 1.04374.6025 1.04374.2511 1.04374.4000
n-Heksan, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
n-Heksan
Formel: H₃C(CH₂)₄CH₃
Kokepunkt: 69 °C (1013 hPa)
Tetthet: 0,659 g/cm³ (20 °C)
Flash punkt: –22 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009520
CAS-nummer: 110-54-3
EINECS: 203-777-6
FN: 1208
ADR: 3,II
Merck Indeks: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) ≥ 96.0 %
Purity Σ hexane isomers + methylcyclopentane (GC) ≥ 98.5 %
Identity (IR) conforms
Color ≤ 10 Hazen
Water-soluble titrable acid ≤ 0.0003 meq/g
Refractive index (n 20/D) 1.375 - 1.376
Density (d 20 °C/20 °C) 0.659 - 0.663
Boiling range (min.95 %) 67 - 69 °C
Transmission (between 260 nm and 420 nm) ≥ 97 %
Thiophene conforms
Aromatics (as benzene) ≤ 0.01 %
Sulfur compounds (as S) ≤ 0.005 %
Readily carbonizable substances conforms
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR