Kaliumklorid 3 mol/l (3 N) in aqueous solution, Supelco®

Leverandør: Merck

Synonymer: Potassiumchloride

M-Clarity™ quality level = MQ100

1.04817.0250
1.04817.0250EA 409.5 NOK
1.04817.0250
Kaliumklorid 3 mol/l (3 N) in aqueous solution, Supelco®
Kaliumklorid
Formel: KCl
Tetthet: 1,13 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8

Limited Time Sale

Select items below are currently on sale

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.13 g/cm3 (20 °C)
pH 5.5 (H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR