Kaliumdihydrogenortofosfat 1/15 mol/l in aqueous solution, Supelco®

Leverandør: Merck

Synonymer: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

M-Clarity™ quality level = MQ100

1.04875.1000
1.04875.1000EA 723 NOK
1.04875.1000
Kaliumdihydrogenortofosfat 1/15 mol/l in aqueous solution, Supelco®
Kaliumdihydrogenortofosfat
Formel: KH₂PO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011401
CAS-nummer: 7778-77-0
EINECS: 231-913-4

Bestill nå

Spesifikasjon Testresultater

Density 1.001 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR