Kaliumbromid, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ300

1.04905.0500
1.04905.0500EA 1075 NOK
1.04905.0500
Kaliumbromid, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Potassium bromide
Formel: KBr
Kokepunkt: 1435 °C (1013 hPa)
Smeltepunkt: 730 °C
Tetthet: 2,75 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011358
CAS-nummer: 7758-02-3
EINECS: 231-830-3
Merck Indeks: 13,07701

Bestill nå

Spesifikasjon Testresultater

Solubility 650 g/l (20 °C)
Melting Point 730 °C
Molar Mass 119.00 g/mol
Bulk Density 900 - 1000 kg/m3
Boiling Point 1435 °C
Vapor Pressure 1.3 hPa (795 °C)
Density 2.75 g/cm3 (20 °C)
pH 5.5 - 8.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR