Kaliumcyanid, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

1.04965.1000
1.04965.1000EA 1480 NOK
1.04965.1000
Kaliumcyanid, Supelco®
Potassium cyanide
Formel: KCN
MW: 65.12 g/mol
Kokepunkt: 1625
Smeltepunkt: 634
Tetthet: 1.55 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011397
CAS-nummer: 151-50-8
EINECS: 205-792-3
FN: 1680
ADR: 6.1 I
Merck Indeks: 13,07709

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR