Kaliumjodat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.05051.0100 1.05051.0500 1.05051.9025
1.05051.0100EA 1090 NOK
1.05051.0100 1.05051.0500 1.05051.9025
Kaliumjodat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Kaliumjodat
Formel: KlO₃
Smeltepunkt: 560 °C
Tetthet: 3,98 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011406
CAS-nummer: 7758-05-6
EINECS: 231-831-9
FN: 1479
ADR: 5.1,II
Merck Indeks: 13,07726

Bestill nå

Spesifikasjon Testresultater

Solubility 92 g/l (25 °C)
Melting Point 560 °C decomposes
Molar Mass 214 g/mol
Bulk Density 2000 kg/m3
Density 3.98 g/cm3 (20 °C)
pH 6 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR