Kaliumnitrit, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.05067.0250
1.05067.0250EA 930 NOK
1.05067.0250
Kaliumnitrit, EMSURE® for analysis, Supelco®
Potassium nitrite
Formel: KNO₂
MW: 85.1 g/mol
Smeltepunkt: 387
Tetthet: 1.92 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011408
CAS-nummer: 7758-09-0
EINECS: 231-832-4
FN: 1488
ADR: 5.1 II
Merck Indeks: 13,07734

Bestill nå

Spesifikasjon Testresultater

Solubility 2810 g/l (20 °C)
Melting Point 440 °C
Molar Mass 85.11 g/mol
Bulk Density 980 kg/m3
Density 1.92 g/cm3 (20 °C)
pH 7 - 10 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR