Potassium pyroantimonate ≥99.0%, krystaller, EMSURE® for analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ300

1.05110.0100
1.05110.0100EA 591 NOK
1.05110.0100
Potassium pyroantimonate ≥99.0%, krystaller, EMSURE® for analysis, Supelco®
Potassium pyroantimonate
Formel: KSbO₃·3H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011415
CAS-nummer: 12208-13-8
EINECS: 235-387-7
FN: 1549
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Identity passes test
Assay (iodometric, calculated on calcined substance) ≥ 99.0 %
Free alkali (as KOH) ≤ 0.1 %
Na (Sodium) ≤ 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR