Kaliumsulfat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ300

1.05153.0500 1.05153.1000 1.05153.5000 1.05153.9025
1.05153.0500EA 593 NOK
1.05153.0500 1.05153.1000 1.05153.5000 1.05153.9025
Kaliumsulfat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Potassium sulphate
Formel: K₂SO₄
Kokepunkt: 1698 °C (1013 hPa)
Smeltepunkt: 1069 °C
Tetthet: 2,66 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011388
CAS-nummer: 7778-80-5
EINECS: 231-915-5
Merck Indeks: 13,07759

Bestill nå

Spesifikasjon Testresultater

Solubility 111 g/l (20 °C)
Melting Point 1069 °C
Molar Mass 174.26 g/mol
Bulk Density 800 kg/m3
Boiling Point 1689 °C (1013 hPa)
Density 2.66 g/cm3 (20 °C)
pH 5.5 - 8.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR