Litiumkarbonat, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Leverandør: Merck
Warning


MQ300

1.05680.0250
1.05680.0250EA 2660 NOK
1.05680.0250
Litiumkarbonat, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
Litiumkarbonat
Formel: Li₂CO₃
MW: 73.89 g/mol
Smeltepunkt: 720
Tetthet: 2.11 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011084
CAS-nummer: 554-13-2
EINECS: 209-062-5
Merck Indeks: 13,05549

Bestill nå

Spesifikasjon Testresultater

Solubility 13 g/l (20 °C)
Melting Point 720 °C
Molar Mass 73.89 g/mol
Bulk Density 250 kg/m3
Vapor Pressure (20 °C)
Density 2.1 g/cm3 (20 °C)
pH 10 - 11 (5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR