Molekylsil 3A (0,3 nm, 3 Å), beads 2 mm, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.05704.0250 1.05704.1000 1.05704.9010
1.05704.0250EA 1280 NOK
1.05704.0250 1.05704.1000 1.05704.9010
Molekylsil 3A (0,3 nm, 3 Å), beads 2 mm, Supelco®
Molekylsil 3A (0,3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131613
CAS-nummer: 308080-99-1
EINECS: 215-283-8

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Bulk Density 700 - 750 kg/m3
Synonyms Sodium aluminium silicate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR