Molekylsil 3A (0,3 nm, 3 Å) with moisture indicator ~10 mesh, Ø ~2 mm, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.05734.0250 1.05734.1000
1.05734.0250EA 1280 NOK
1.05734.0250 1.05734.1000
Molekylsil 3A (0,3 nm, 3 Å) with moisture indicator ~10 mesh, Ø ~2 mm, Supelco®
Molekylsil 3A (0,3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL nummer: MFCD00131613
CAS-nummer: 308080-99-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR