Molekylsil 3A (0,3 nm, 3 Å), staver, Ø ~1,6 mm, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.05741.1000 1.05741.0250
1.05741.1000EA 2810 NOK
1.05741.1000 1.05741.0250
Molekylsil 3A (0,3 nm, 3 Å), staver, Ø ~1,6 mm, Supelco®
Molekylsil 3A (0,3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131613
CAS-nummer: 308080-99-1
EINECS: 215-283-8

Bestill nå

Spesifikasjon Testresultater

Identity Conforms
Loss on ignition (950 °C) ≤10 %
Water absorption capacity (24 hrs.,80 % relative humidity) ≥15%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR