Magnesium ≥99.5%, folie, Width: 3 mm, Thickness: 0.15-0.30 mm, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

1.05812.0001_U 1.05812.0001
MERC1.05812.0001_UEA 0 NOK
MERC1.05812.0001_U 1.05812.0001
Magnesium ≥99.5%, folie, Width: 3 mm, Thickness: 0.15-0.30 mm, Supelco®
Magnesium
Formel: Mg
Kokepunkt: 1107 °C (1013 hPa)
Smeltepunkt: 651 °C
Tetthet: 1,74 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085308
CAS-nummer: 7439-95-4
EINECS: 231-104-6
FN: 1869
ADR: 4.1,III
Merck Indeks: 13,05675

Bestill nå

Spesifikasjon Testresultater

Assay (complexometric) ≥ 99.5 %
Substances insoluble in hydrochloric acid ≤ 0.05 %
Cu (Copper) ≤ 0.005 %
Fe (Iron) ≤ 0.05 %
Pb (Lead) ≤ 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR