Magnesium nitrate (≥10,0 ± 0,2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.05813.0050
1.05813.0050EA 1470 NOK
1.05813.0050
Magnesium nitrate (≥10,0 ± 0,2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Magnesium nitrate solutions

Magnesium matrix modifier for graphite furnace AAS.

Formel: Mg(NO₃)₂
Tetthet: 1,05 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
CAS-nummer: 10377-60-3
FN: 2031
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Ag (Silver) ≤ 0.000002 g/l
As (Arsenic) ≤ 0.00001 g/l
Cd (Cadmium) ≤ 0.000001 g/l
Pb (Lead) ≤ 0.00001 g/l
Se (Selenium) ≤ 0.0001 g/l
Tl (Thallium) ≤ 0.00001 g/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR