Magnesium ≥98.5%, pulver, Ø ~ 0.06-0.3 mm, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

1.05815.1000
1.05815.1000EA 2300 NOK
1.05815.1000
Magnesium ≥98.5%, pulver, Ø ~ 0.06-0.3 mm, Supelco®
Magnesium
Formel: Mg
Kokepunkt: 1107 °C (1013 hPa)
Smeltepunkt: 651 °C
Tetthet: 1,74 g/cm³ (25 °C)
Flash punkt: 500 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085308
CAS-nummer: 7439-95-4
EINECS: 231-104-6
FN: 1418
ADR: 4.3 (4.2),II

Bestill nå

Spesifikasjon Testresultater

Assay (complexometric) ≥ 98.5 %
Substances insoluble in hydrochloric acid ≤ 0.05 %
Fe (Iron) ≤ 500 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR