Magnesiumnitrat heksahydrat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ300

1.05853.0500 1.05853.9025
1.05853.0500EA 754 NOK
1.05853.0500 1.05853.9025
Magnesiumnitrat heksahydrat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Magnesiumnitrat heksahydrat
Formel: Mg(NO₃)₂·6H₂O
Kokepunkt: 330 °C (1013 hPa)
Smeltepunkt: 89 °C
Tetthet: 1,63 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149779
CAS-nummer: 13446-18-9
EINECS: 233-826-7
FN: 1474
ADR: 5.1,III
Merck Indeks: 13,05697

Bestill nå

Spesifikasjon Testresultater

Solubility 420 g/l (20 °C)
Melting Point 89 °C (for the hexahydrate)
Molar Mass 256.41 g/mol
Density 1.46 g/cm3 (20 °C)
pH 5.0 - 7.0 (50 g/l, H2O, 25 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR