Diklormetan stabilised, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Warning


Stabilisering: stabilisert med 2-metyl-2-buten 20 ppm
M-Clarity™ quality level = MQ300

1.06050.1000 1.06050.2500 1.06050.6010 1.06050.6025 1.06050.9190 1.06050.4000
1.06050.1000EA 576 NOK
1.06050.1000 1.06050.2500 1.06050.6010 1.06050.6025 1.06050.9190 1.06050.4000
Diklormetan stabilised, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Diklormetan

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: CH₂Cl₂
Kokepunkt: 39,8 °C (1013 hPa)
Smeltepunkt: –95 °C
Tetthet: 1,322 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
FN: 1593
ADR: 6.1,III
Merck Indeks: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 20 g/l (20 °C)
Melting Point -95 °C
Molar Mass 84.93 g/mol
Boiling Point 40 °C (1013 hPa)
Vapor Pressure 475 hPa (20 °C)
Refractive Index 1.42
Explosion Limit 13 - 22 %(V)
Density 1.33 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 605 °C DIN 51794
Evaporation number 1.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR