Metylrødt, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.06076.0025 1.06076.0100
1.06076.0025EA 698 NOK
1.06076.0025 1.06076.0100
Metylrødt, Supelco®
Metylrødt
Formel: C₁₅H₁₅N₃O₂
Smeltepunkt: 178…182 °C
Tetthet: 1,31 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002425
CAS-nummer: 493-52-7
EINECS: 207-776-1
Merck Indeks: 12,06199

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) slightly soluble
Melting Point 178 - 182 °C
Molar Mass 269.31 g/mol
Bulk Density 300 - 500 kg/m3
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR